Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Fluoroindoline, 97%

Catalog No. AAH5415806
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5415806 5 g
AAH54158MD 250 mg
AAH5415803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH5415806 Supplier Thermo Scientific Chemicals Supplier No. H5415806
Only null left
Add to Cart

CAS: 2343-22-8 | C8H8FN | 137.157 g/mol

5-Fluoroindoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Fluoroindoline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2343-22-8
Molecular Formula C8H8FN
Molecular Weight (g/mol) 137.157
MDL Number MFCD00214461
InChI Key NXQRMQIYCWFDGP-UHFFFAOYSA-N
PubChem CID 2774463
IUPAC Name 5-fluoro-2,3-dihydro-1H-indole
SMILES C1CNC2=C1C=C(C=C2)F

Specifications

Quantity 5 g
Sensitivity Air sensitive
Synonym 5-fluoroindoline, 5-fluoro-2,3-dihydro-1h-indole, 1h-indole,5-fluoro-2,3-dihydro, 1h-indole, 5-fluoro-2,3-dihydro, 5-fluoro-indoline, peakdale1_000398, 5-fluoro-2,3-dihydro-1h-indol, 2,3-dihydro-5-fluoro-1h-indole
Solubility Information Slightly soluble in water.
Formula Weight 137.16
Percent Purity 97%
Chemical Name or Material 5-Fluoroindoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.