Learn More
Thermo Scientific Chemicals 5-Fluorouracil, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• 5-Fluorouracil is an antimetabolite and antineoplastic that prevents DNA synthesis by blocking conversion of deoxyuridylic acid
• 5-Fluorouracil can replace uracil in RNA and prevent protein synthesis and cell growth
Applications
• 5-Fluorouracil can be used for laboratory research on tumor growth and immunosuppression
• 5-Fluorouracil may also be appropriate for laboratory studies of neurotoxicity, cell lysis, and DNA damage
Specifications
Specifications
| Melting Point | 278.0°C to 282.0°C |
| Color | White |
| Boiling Point | 190.0°C to 200.0°C (0.1 mmHg) |
| Loss on Drying | 0.5% (80°C, 4 hrs) (in vacuum) max. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Glass bottle |
| Quantity | 100 g |
| Beilstein | 24,IV,1229 |
| Merck Index | 15,4213 |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement Suspected of causing genetic defects. Suspected of damaging the unborn child. Toxic if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes serious eye irritation. Causes skin irri |
| Precautionary Statement | GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. IF INHALED: Remove victim to fresh air and keep at rest in a position comforta |
| EINECSNumber | 200-085-6 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.