Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Formylcytosine (5-fC) Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501993248
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3248 100 μg
50-199-3249 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3248 Supplier Active Motif Supplier No. 61227
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

5-formylcytosine is an oxidized derivative of 5-methylcytosine, a type of DNA methylation. 5-formylcytosine is a nucleobase analogue with a cytosine that possesses a hydrogen at position 5 that has been replaced by a formyl group. 5-methylcytosine is oxidized to 5-hydroxymethylcytosine which is then oxidized to 5-formylcytosine and later to 5-carboxylcytosine. These oxidations are catalyzed by the Ten-Eleven Translocation (TET) enzyme family. 5-formylcytosine has been shown to determine nucleosome organization and plays a role in regulating transcription.

Specifications

Antigen 5-Formylcytosine (5-fC)
Applications Dot Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Host Species Rabbit
Immunogen This 5-Formylcytosine antibody was raised against 5-formylcytidine conjugated to kLH and recognizes 5-formylcytosine.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.