Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Formylcytosine (5-fC) Rabbit anti-Chemical, Unconjugated, Polyclonal, Active Motif™
SDP

Catalog No. 501994145
Encompass
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4145 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-4145 Supplier Active Motif Supplier No. 61223
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

5-formylcytosine is an oxidized derivative of 5-methylcytosine, a type of DNA methylation. 5-formylcytosine is a nucleobase analogue with a cytosine that possesses a hydrogen at position 5 that has been replaced by a formyl group. 5-methylcytosine is oxidized to 5-hydroxymethylcytosine which is then oxidized to 5-formylcytosine and later to 5-carboxylcytosine. These oxidations are catalyzed by the Ten-Eleven Translocation (TET) enzyme family. 5-formylcytosine has been shown to determine nucleosome organization and plays a role in regulating transcription.

Specifications

Antigen 5-Formylcytosine (5-fC)
Applications Dot Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Host Species Rabbit
Immunogen This 5-Carboxylcytosine antibody was raised against 5-carboxylcytidine conjugated to kLH and recognizes 5-carboxylcytosine.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.