Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Formylcytosines Antibody Pair, Biotin, R&D Systems™
SDP

Catalog No. NB287462 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB287462 1 Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB287462 Supplier R&D Systems Supplier No. NBP323469

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

A Novus product, part of R&D Systems. Rabbit Monoclonal Antibody

5-Formylcytosines Monoclonal specifically detects 5-Formylcytosines in Human samples. It is validated for ELISA, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair).

Specifications

Antigen 5-Formylcytosines
Applications ELISA, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
Classification Monoclonal
Conjugate Biotin
Dilution ELISA, ELISA Capture (Matched Antibody Pair) 50 - 200ng/well, ELISA Detection (Matched Antibody Pair) 0.1 - 0.5ug/ml
Host Species Rabbit
Quantity 1 Set
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.