Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Labconco 5 ft. Nexus Horizontal Clean Bench
GSA_VASDP

Catalog No. 17194966 Shop All Labconco Corporation Products
Change view
Click to view available options
Material:
Epoxy-coated Steel
Stainless Steel
Voltage:
115 V
Includes:
Base Stand
Clean Bench Only
Clean Bench and Base Stand
UV Lamp and Base Stand
Depth (English) Interior:
21 in.
26 in.
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material Voltage Includes Depth (English) Interior
17-194-966 Epoxy-coated Steel 115 V Clean Bench Only 26 in.
17-194-943 Stainless Steel 115 V Clean Bench Only 21 in.
17-195-098 Stainless Steel 115 V Clean Bench and Base Stand 21 in.
17-194-973 Stainless Steel 115 V UV Lamp and Base Stand 21 in.
17-195-014 Stainless Steel 115 V Clean Bench Only 26 in.
17-194-975 Stainless Steel 115 V Base Stand 26 in.
17-194-992 Stainless Steel 115 V UV Lamp and Base Stand 26 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-194-966 Supplier Labconco Supplier No. 5351000
Add to Cart
Add to Cart
Request a Quote

Nexus Horizontal Clean Benches are designed to protect non-hazardous applications by providing an ISO Class 4 clean airflow environment. The large, 5 in. touchscreen interface features Nexus OS™ software that simplifies operation, maintains records and provides user customizaton of the system.

  • 5 ft. Nexus Horizontal Clean Bench
  • ISO Class 4 laminar airflow is 10 times cleaner than the industry standard ISO Class 5
  • Energy efficient ECM motor with CAP™ Technology ensures accurate airflow between certifications

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Product Type Horizontal Clean Bench
Coating Epoxy
Filter Type HEPA
Lighting Type LED
Material Epoxy-coated Steel
Color Optic White
Depth (English) Interior 26 in.
Depth (Metric) Interior 66.6 cm
Height (English) 50.1 in.
Height (English) Interior 30.2 in.
Height (Metric) 127.5 cm
Height (Metric) Interior 76.7 cm
Includes Clean Bench Only
IP Rating None
Plug Type NEMA 5-20P
Voltage 115 V
Width (English) 63.4 in.
Width (English) Interior 61.0 in.
Width (Metric) 161.0 cm
Width (Metric) Interior 155.0 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.