Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hexen-1-ol, 99%

Catalog No. p-4509449
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle

CAS: 821-41-0 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 821-41-0
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
InChI Key UIZVMOZAXAMASY-UHFFFAOYSA-N
Synonym 5-hexen-1-ol, 1-hexen-6-ol, 5-hexenol, unii-57pd1rf6g7, 5-hexene-1-ol, 5-hexanol, hex-1-en-6-ol, acmc-209pn9, ho ch2 4ch=ch2
PubChem CID 69963
IUPAC Name hex-5-en-1-ol
SMILES C=CCCCCO

Specifications

Color Colorless
Density 0.8725g/mL
Boiling Point 152.0°C to 155.0°C
Flash Point 47°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4340 to 1.4360
Linear Formula H2C=CH(CH2)4OH
Quantity 5 mL
Beilstein 01,IV,2137
Specific Gravity 0.8725
Solubility Information Solubility in water: 18.6g/L (20°C).
Formula Weight 100.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 5-Hexen-1-ol
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 212-477-4

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.