Learn More
5-Hexyn-1-ol, 97%, Thermo Scientific Chemicals
Description
5-Hexyn-1-ol is used as starting material for the preparation of terminal alkynes and lactones containing terminal alykyne groups. It is also used in the synthesis of cinnoline-fused cyclic enediyne, llycopodium alkaloids, (+)-nankakurine A and (+)-nankakurine B and 7-benzoyloxy-3-(2-nitrophenylseleno)-1,5-cyclodecadiyne.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | Colorless to Yellow |
Density | 0.880 g/mL |
Boiling Point | 73°C to 75°C (15 mmHg) |
Flash Point | 70°C (158°F) |
Odor | Characteristic |
Refractive Index | 1.4451 |
Linear Formula | HC≡C(CH2)3CH2OH |
Quantity | 5 g |
Solubility Information | Slightly miscible with water. |
Formula Weight | 98.15 |
Show More |
Safety and Handling
Hazard Category | H227-H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335-H227 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Combustible liquid. |
Precautionary Statement | P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.