Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Potassium hydroxide, 1N solution in ethanol

Catalog No. AC393322500
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Plastic bottle
This item is not returnable. View return policy

Catalog No. AC393322500

Supplier: Thermo Scientific Chemicals 393322500

Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 1310-58-3 | HKO | 56.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Potassium hydroxide
Color Brown-Orange to Yellow
Name Note 1N solution in ethanol
Boiling Point 78.0°C
CAS 1310-58-3, 64-17-5
Melting Point -117.0°C
Molecular Formula HKO
Linear Formula KOH
Concentration 0.950 to 1.050N
MDL Number MFCD00003553
Density 0.8600g/mL
Merck Index 15, 7761
Synonym potassium hydroxide, caustic potash, potash lye, potassium hydrate, hydroxyde de potassium, potassa, potasse caustique, potassium hydroxide k oh, potassium hydroxide solution, caswell no. 693
Solubility Information Solubility in water: soluble.
Quantity 250 mL
InChI Key KWYUFKZDYYNOTN-UHFFFAOYSA-M
SMILES [OH-].[K+]
IUPAC Name potassium hydroxide
Molecular Weight (g/mol) 56.11
PubChem CID 14797
Flash Point 12°C
ChEBI CHEBI:32035
Formula Weight 56.11
Packaging Plastic bottle
Specific Gravity 0.86
Physical Form Liquid
Show More Show Less

For Manufacturing and Laboratory Use. Not for beverage use

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.