Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-hmC Antibody (AB3/63.3), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP250099B
Description
5-hmC Monoclonal specifically detects 5-hmC in All Species samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
5-hmC | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
HMCES | |
5-hmC conjugated to BSA | |
0.1 mL | |
DNA Repair, Epigenetics, Stem Cells | |
56941 | |
All species | |
Purified |
Immunocytochemistry, Immunofluorescence | |
AB3/63.3 | |
Immunocytochemistry/Immunofluorescence | |
5'-hmC, 5-hydroxymethylcytosine | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Expected to detect 5-hydroxymethylcytosine in all species. | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction