Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-hmC Antibody (C.15220001), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP259178
Description
5-hmC Monoclonal specifically detects 5-hmC in Human, Mouse, Drosophila samples. It is validated for ELISA, Dot Blot, Methylated DNA Immunoprecipitation.Specifications
5-hmC | |
Monoclonal | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
HMCES | |
The exact immunogen is propietary information. | |
100 μg | |
DNA Repair, Epigenetics, Stem Cells | |
56941 | |
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
ChIP Assay, Dot Blot | |
C.15220001 | |
ELISA 1:1000, Dot Blot 1:500, Methylated DNA Immunoprecipitation 2.5 μg/IP | |
5'-hmC, 5-hydroxymethylcytosine | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Drosophila | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction