Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HT1B Antibody, Novus Biologicals™
SDP

Catalog No. NB10056350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10056350 0.1 mg
NB10056350T 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10056350 Supplier Novus Biologicals Supplier No. NB10056350
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 1 publication

5-HT1B Polyclonal specifically detects 5-HT1B in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen 5-HT1B
Applications Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml
Formulation PBS with 0.05% Sodium Azide
Gene Accession No. P28222, P28334, P28564
Gene Alias 5-HT1B5-HT-1B, 5-HT1DB, 5-hydroxytryptamine (serotonin) receptor 1B, HTR1D2,5-hydroxytryptamine receptor 1B, HTR1DB, S125-HT-1D-beta, Serotonin 1D beta receptor, Serotonin receptor 1B
Gene Symbols HTR1B
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 8-26 and 263-278 of rat 5-HT1BR (Uniprot# P28564)
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3351
Test Specificity A 40-41 kDa band is detected.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.