Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-HT3B Antibody (2H9H5) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$548.00
Specifications
Antigen | 5-HT3B |
---|---|
Clone | 2H9H5 |
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500 |
Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP2-61787
![]() |
Novus Biologicals
NBP261787 |
0.1 mL |
Each of 1 for $548.00
|
|
|||||
Description
5-HT3B Monoclonal specifically detects 5-HT3B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
5-HT3B | |
1 mg/mL | |
Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
Mouse | |
5-HT3-B, 5-HT3B5-hydroxytryptamine receptor 3B, 5-hydroxytryptamine (serotonin) receptor 3B, 5-hydroxytryptamine 3 receptor B subunit, Serotonin receptor 3B, serotonin-gated ion channel subunit | |
HTR3B | |
IgG2b | |
Protein G purified |
2H9H5 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
Monoclonal | |
Purified | |
Signal Transduction | |
9177 | |
Purified recombinant fragment of human HTR3B (AA: extra 23-150) expressed in E. Coli. | |
Primary | |
50.3 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title