Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-HT4 Antibody, DyLight 650, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178403C
Description
5-HT4 Polyclonal specifically detects 5-HT4 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
5-HT4 | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Q13639 | |
HTR4 | |
A synthetic peptide made to a C-terminal portion of the human 5HT4 Receptor protein (between residues 350-388) [UniProt Q13639, isoform 1] | |
0.1 mL | |
Asthma, GPCR, Growth and Development, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
3360 | |
Store at 4C in the dark. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
5-HT-4, 5-HT4R, 5-hydroxytryptamine (serotonin) receptor 4,5-hydroxytryptamine receptor 4,5-HT45-hydroxytryptamine4 receptor, cardiac 5-HT4 receptor, Serotonin receptor 4 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction