Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-HT6 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP310802100UL
Description
5-HT6 Polyclonal specifically detects 5-HT6 in Human samples. It is validated for Western Blot.Specifications
5-HT6 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
5-hydroxytryptamine (serotonin) receptor 6,5-HT65-HT6R, Serotonin receptor 6,5-HT-6,5-hydroxytryptamine receptor 6 | |
The immunogen is a synthetic peptide directed towards the C terminal region of human 5-HT6 (NP_000862.1). Peptide sequence PDSDSDSDAGSGGSSGLRLTAQLLLPGEATQDPPLPTRAAAAVNFFNIDP | |
100 μg | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
3362 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction