Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-HT7 Antibody (SR1954), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP322298100UL
Description
5-HT7 Monoclonal antibody specifically detects 5-HT7 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
5-HT7 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot | |
SR1954 | |
Western Blot 1:500-1:2000 | |
5-HT-7, 5-HT75-hydroxytryptamine receptor 7, 5-HT-X, 5-hydroxytryptamine (serotonin) receptor 7 (adenylate cyclase-coupled), serotonin 5-HT-7 receptor, Serotonin receptor 7 | |
A synthesized peptide derived from human 5-HT7 (Uniprot #: P34969) | |
100 μg | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
3363 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction