Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-HTR1A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981206
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1206 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1206 Supplier Bioss Supplier No. BS1124R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This is one of the several different receptors for 5-hydroxytryptamine (serotonin), a biogenic hormone that functions as a neurotransmitter, a hormone, and a mitogen. The activity of this receptor is mediated by G proteins that inhibit adenylate cyclase activity.

Specifications

Antigen 5-HTR1A
Applications Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene HTR1A
Gene Accession No. P08908
Gene Alias 5ht1a; 5-HT1A; 5-HT-1A; 5-HT1A receptor; 5-hydroxytryptamine (serotonin) receptor 1A; 5-hydroxytryptamine (serotonin) receptor 1A, G protein-coupled; 5-hydroxytryptamine receptor 1A; ADRB2RL1; ADRBRL1; G-21; Gpcr18; G-protein coupled receptor 18; guanine nucleotide-binding regulatory protein-coupled receptor; HTR1A; PFMCD; RAT5HT1A; Serotonin receptor 1A
Gene Symbols HTR1A
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from rat 5-HTR1A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3350
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.