Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

phyproof™ 5-Hydroxy-L-tryptophan, ≥98% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111002103 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 mg
Catalog No. Quantity
11-100-2103 50 mg
1 options

Catalog No. 11-100-2103

Supplier: MilliporeSigma Supelco PHL8047750MG

Only null left
Add to Cart
Add to Cart

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

TRUSTED_SUSTAINABILITY

Specifications

CAS 9-8-4350
Product Line phyproof
Density 0.902 g/mL (at 25°C)
Linear Formula C11H12N2O3
MDL Number MFCD00064341
Melting Point 270°C
Type Reference Substance
Quantity 50 mg
Formula Weight 220.22
Percent Purity ≥98% (HPLC)

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.