Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxyindan 99.0+%, TCI America™
SDP

Catalog No. H0251100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0251100G 100 g
H025125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H0251100G Supplier TCI America Supplier No. H0251100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1470-94-6
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.18
MDL Number MFCD00003802
InChI Key PEHSSTUGJUBZBI-UHFFFAOYSA-N
Synonym 5-indanol, indan-5-ol, 5-hydroxyindan, 5-hydroxyhydrindene, 5-hydroxyindane, 1h-inden-5-ol, 2,3-dihydro, indanol-5, unii-9z94h6f15t, 5-indano
PubChem CID 15118
ChEBI CHEBI:59311
IUPAC Name 2,3-dihydro-1H-inden-5-ol
SMILES OC1=CC=C2CCCC2=C1

Specifications

Melting Point 53°C
Color White-Yellow
Quantity 100 g
UN Number 2811
Formula Weight 134.18
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxyindan
RTECSNumber NK7525000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.