Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxyindole, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
5g
Packaging:
Glass bottle
1953-54-4
C8H7NO
133.15
MFCD00005677
LMIQERWZRIFWNZ-UHFFFAOYSA-N
5-hydroxyindole,indol-5-ol,5-hydroxy-1h-indole,hydroxy-5 indole,5-hydroxy-indole,5-indolol,hydroxy-5-indole,5-hydroxyindol,5-hydroxy indole,unii-320un7xzyn
16054
1H-indol-5-ol
C1=CC2=C(C=CN2)C=C1O
1953-54-4
C8H7NO
133.15
MFCD00005677
LMIQERWZRIFWNZ-UHFFFAOYSA-N
5-hydroxyindole,indol-5-ol,5-hydroxy-1h-indole,hydroxy-5 indole,5-hydroxy-indole,5-indolol,hydroxy-5-indole,5-hydroxyindol,5-hydroxy indole,unii-320un7xzyn
16054
1H-indol-5-ol
C1=CC2=C(C=CN2)C=C1O
Quantity 250mg
Infrared Spectrum Authentic
Melting Point 105.0°C to 110.0°C
Color Beige to Brown
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Beilstein 21, III, 765
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in acetone,ethanol and acetic acid
Formula Weight 133.15
Physical Form Crystalline Needles or Powder
Percent Purity 97%
Chemical Name or Material 5-Hydroxyindole
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.