Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-(Hydroxymethyl)furfural, 98%

Catalog No. AC121460250
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 67-47-0 | C6H6O3 | 126.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-47-0
Molecular Formula C6H6O3
Molecular Weight (g/mol) 126.11
InChI Key NOEGNKMFWQHSLB-UHFFFAOYSA-N
Synonym 5-hydroxymethylfurfural, 5-hydroxymethyl-2-furaldehyde, 5-hydroxymethyl furfural, 5-hydroxymethyl furan-2-carbaldehyde, hydroxymethylfurfural, 5-hydroxymethyl-2-furfural, 5-oxymethylfurfurole, 5-hydroxymethylfuraldehyde, hydroxymethylfurfurole
PubChem CID 237332
ChEBI CHEBI:412516
IUPAC Name 5-(hydroxymethyl)furan-2-carbaldehyde
SMILES C1=C(OC(=C1)C=O)CO

Specifications

Melting Point 30.0°C to 34.0°C
Color Yellow
Boiling Point 114°C to 116°C (1.0 mmHg)
Flash Point 79°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5627
Quantity 25 g
Beilstein 18, V,1, 130
Merck Index 15,4871
Solubility Information Solubility in water: soluble. Other solubilities: soluble in most common organic solvents
Formula Weight 126.11
Percent Purity 98%
Physical Form Crystalline Powder, Chunks or Liquid
Chemical Name or Material 5-(Hydroxymethyl)furfural
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 200-654-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.