Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxymethylcytosine (5-hmC) Monoclonal Antibody (59.1), Active Motif™
SDP

Mouse Monoclonal Antibody

$147.60 - $584.25

Specifications

Antigen 5-Hydroxymethylcytosine (5-hmC)
Clone 59.1
Concentration 1 mg/mL
Applications ChIP Assay, Dot Blot, Immunoprecipitation
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3030
SDP
Encompass
View Documents
Active Motif
40000
10 μg
Each for $147.60
Only null left
Add to Cart
 
50-199-3031
SDP
Encompass
View Documents
Active Motif
40900
50 μg
Each for $332.10
Only null left
Add to Cart
 
50-199-3029
SDP
Encompass
View Documents
Active Motif
39999
100 μg
Each for $584.25
Only null left
Add to Cart
 
Description

Description

5-hydroxymethylcytosine (5-hmC) has been recently discovered in mammalian DNA. 5-hmC results from the enzymatic conversion of 5-methylcytosine into 5-hydroxymethylcytosine by the TET family of oxygenases. Initially, the 5-hmC base had been identified in Purkinje neurons, in granule cells and embryonic stem cells where it is present at high levels (up to 0,6% of total nucleotides in Purkinje cells). A recent report indicates that 5-hmC is also abundant in brain tissue, especially in areas that are associated with higher cognitive functions.Early evidence suggests that 5-hmC may represent an intermediate in a new pathway which demethylates DNA, converting 5-mC to cytosine. Although its precise role has still to be shown, recent results indicate, however, that 5-hmC may play important roles distinct from 5-mC. This may open up entirely new perspectives in epigenetic studies.
Specifications

Specifications

5-Hydroxymethylcytosine (5-hmC)
1 mg/mL
Monoclonal
Liquid
RUO
70mM tris with 0.07mM EDTA, 105mM NaCl, 30% glycerol, 31mM glycine and 0.035% sodium azide; pH 8
This 5-Hydroxymethylcytosine antibody was raised against 5-hydroxymethylcytidine conjugated to kLH and recognizes 5-hydroxymethylcytosine.
Primary
-20°C, Avoid Freeze/Thaw Cycles
59.1
ChIP Assay, Dot Blot, Immunoprecipitation
Unconjugated
Mouse
Human, Mouse
5hmc; 5-hmc; 5-HydroxymethylCytidine; C5H7N3O2
IgG2a
Protein G
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.