Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Hydroxymethylcytosine (5-hmC) Polyclonal Antibody, Active Motif™
SDP

Catalog No. 501994115
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
50 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4115 50 μL
50-199-4114 10 μL
50-199-4113 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-199-4115 Supplier Active Motif Supplier No. 39069
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

5-hydroxymethylcytosine (5-hmC) has been recently discovered in mammalian DNA. 5-hmC results from the enzymatic conversion of 5-methylcytosine into 5-hydroxymethylcytosine by the TET family of oxygenases. Initially, the 5-hmC base had been identified in Purkinje neurons, in granule cells and embryonic stem cells where it is present at high levels (up to 0,6% of total nucleotides in Purkinje cells). A recent report indicates that 5-hmC is also abundant in brain tissue, especially in areas that are associated with higher cognitive functions.Early evidence suggests that 5-hmC may represent an intermediate in a new pathway which demethylates DNA, converting 5-mC to cytosine. Although its precise role has still to be shown, recent results indicate, however, that 5-hmC may play important roles distinct from 5-mC. This may open up entirely new perspectives in epigenetic studies.

Specifications

Antigen 5-Hydroxymethylcytosine (5-hmC)
Applications Dot Blot, Immunoprecipitation
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene Alias 5hmc; 5-hmc; 5-HydroxymethylCytidine; C5H7N3O2
Host Species Rabbit
Immunogen This 5-Hydroxymethylcytosine antibody was raised against 5-hydroxymethylcytidine conjugated to kLH and recognizes 5-hydroxymethylcytosine.
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.