Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-hydroxymethylcytosine (5-hmC) Rabbit, Clone: RM236, Abnova™

Catalog No. 89035324 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-035-324 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-035-324 Supplier Abnova Corporation Supplier No. MAB15116
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Rabbit monoclonal antibody raised against 5-hydroxymethylcytosine (5-hmC).

  • High specificity
  • High reactivity

Specifications

Antigen 5-hydroxymethylcytosine (5-hmC)
Applications ChIP Assay, Dot Blot, ELISA, Immunocytochemistry, Immunohistochemistry (PFA fixed)
Classification Monoclonal
Clone RM236
Conjugate Unconjugated
Description Rabbit monoclonal antibody raised against 5-hydroxymethylcytosine (5-hmC).
Dilution Dot Blot (0.2-1 ug/mL) ELISA (0.1-1 ug/mL) Immunocytochemistry (0.5-2 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (0.1-1 ug/mL) Hydroxymethylated DNA immunoprecipitation (0.2-2 ug/mL) The optimal working dilu
Formulation In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
Host Species Rabbit
Immunogen 5-hydroxymethylcytosine conjugated with BSA.
Purification Method Protein A affinity purification
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity This antibody reacts to 5-hydroxymethylcytosine in both single-stranded and double-stranded DNA. No cross reactivity with non-methylated cytosine and methylcytosine in DNA.
Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.