Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ 5-Hydroxymethylcytosine (5hmC), Mouse, Unlabeled, Clone: EDL HMC 1A,

Catalog No. MABE1093MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MABE1093MI 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE1093MI Supplier MilliporeSigma™ Supplier No. MABE1093

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

5-Hydroxymethylcytosine (also known as 5-hmC) is the first oxidative product in the active demethylation of 5-methylcytosine (5mC). 5hmC is a DNA pyrimidine nitrogen base and is a predominantly stable DNA modification. It is produced by the action of three Ten-eleven translocation 1 (TET1) enzyme on 5mC. Each mammalian cell contains 5hmC, but its levels vary significantly depending on the cell type. Higher levels of 5hmC are found in neuronal cells and its level is shown to increase with age in hippocampus and cerebellum. It is also prevalent in embryonic stems cells and its reduced levels are shown to result in impaired self-renewal of stem cells. In the brain and in embryonic stem cells, 5hmC in enriched in promoters, gene bodies, and intergenic areas near genes and positively correlates with gene expression at these loci. 5hmC appears to play a role to promote gene expression during active demethylation. Significantly lower of 5hmC have been reported in cancer cell lines, melanoma, adenomas, and carcinomas compared to the normal surrounding tissue.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5-Hydroxymethylcytosine (5hmC)
Applications Immunoprecipitation
Classification Monoclonal
Clone EDL HMC 1A
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Formulation Purified mouse IgG2b in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% Sodium Azide.
Host Species Mouse
Immunogen BSA-conjugated 5-hydroxymethylcytidine.
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Test Specificity Clone EDL HMC 1A targets 5-hydroxymethylcytosine (5hmC), but not 5-methylcytosine (5mC), 5-carboxylcytosine (5caC), 5-formylcytosine (5fC), or unmodified cytosine (C).
Target Species All species
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.