Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 5-Hydroxytryptophan Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5119514
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5119514 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5119514 Supplier Invitrogen™ Supplier No. PA5119514
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.

5-Hydroxytryptophan (5-HTP) is an intermediate metabolite in the biochemical synthesis of serotonin from the essential amino acid tryptophan. The conversion of tryptophan to 5-HTP involves the enzyme tryptophan hydroxylase (TPH), which exists in two isoforms: TPH1 and TPH2. TPH1 is mainly expressed in the peripheral tissues, while TPH2 is predominantly found in the central nervous system. Once synthesized, 5-HTP is further decarboxylated by aromatic L-amino acid decarboxylase (AADC) to produce serotonin (5-hydroxytryptamine, 5-HT). This biogenic amine plays a crucial role in regulating mood, cognition, and various physiological processes. Serotonin can also be acetylated and methylated to produce melatonin, a hormone involved in regulating sleep-wake cycles. Given its central role in serotonin biosynthesis, 5-HTP is often used in the treatment of conditions like depression, anxiety, and insomnia. Recent biotechnological advances have focused on optimizing the production of 5-HTP through engineered enzyme systems to meet increasing demand.
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5-Hydroxytryptophan
Applications Dot Blot, ELISA
Classification Polyclonal
Concentration 0.63 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.025% ProClin 300; pH 7
Gene Alias 5-HTP; oxitriptan
Host Species Rabbit
Immunogen 5-Hydroxytryptophan.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.