Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxy-2-methylbenzselenazole 97%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass Bottle in Plastic Container
2946-17-0
C9H9NOSe
226.148
MFCD00005558
LVIYTMFDCXRSDL-UHFFFAOYSA-N
5-methoxy-2-methylbenzoselenazole,benzoselenazole, 5-methoxy-2-methyl,5-methoxy-2-methylbenzselenazole,2-methyl-5-methoxybenzoselenazole,5-methoxy-2-methylbenzo d 1,3-selenazole,#,5-methoxy-2-methylbenzo d 1,3 selenazole,5-methoxy-2-methylbenzo d 1,3-selenazoline
76274
5-methoxy-2-methyl-1,3-benzoselenazole
CC1=NC2=C([Se]1)C=CC(=C2)OC
2946-17-0
C9H9NOSe
226.148
MFCD00005558
LVIYTMFDCXRSDL-UHFFFAOYSA-N
5-methoxy-2-methylbenzoselenazole,benzoselenazole, 5-methoxy-2-methyl,5-methoxy-2-methylbenzselenazole,2-methyl-5-methoxybenzoselenazole,5-methoxy-2-methylbenzo d 1,3-selenazole,#,5-methoxy-2-methylbenzo d 1,3 selenazole,5-methoxy-2-methylbenzo d 1,3-selenazoline
76274
5-methoxy-2-methyl-1,3-benzoselenazole
CC1=NC2=C([Se]1)C=CC(=C2)OC
Refractive Index 1.6360 to 1.6390 (20°C, 589nm)
Density 1.4900g/mL
Infrared Spectrum Authentic
Packaging Glass Bottle in Plastic Container
Quantity 1g
Color Orange-Red
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 226.14
Physical Form Liquid
Percent Purity ≥96.0%
Chemical Name or Material 5-Methoxy-2-methylbenzselenazole, 97%
Show More Show Less

GHS H Statement:
Toxic if inhaled.
Toxic if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Very toxic to aquatic life with long lasting effects.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger