Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dichloromethane, 99.8%, Extra Dry over Molecular Sieve, Stabilized, AcroSeal™

Catalog No. AC348465000
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
Packaging:
AcroSeal™ Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Dichloromethane, 99.8%, CH2Cl2, CAS Number-75-09-2, 513-35-9, aerothene mm, freon 30, methane dichloride, methane, dichloro, methylene bichloride, methylene chloride, methylene dichloride, narkotil, solaesthin, solmethine, 1L, 01, 60, 39.0 deg.C to 40.0 deg.C, CHEBI:15767, Colorless, >120 deg.C | CAS: 75-09-2 | CH2Cl2 | 84.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-09-2, 513-35-9
Molecular Formula CH2Cl2
Molecular Weight (g/mol) 84.93
MDL Number MFCD00000881
InChI Key YMWUJEATGCHHMB-UHFFFAOYSA-N
Synonym methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
PubChem CID 6344
ChEBI CHEBI:15767
IUPAC Name dichloromethane
SMILES ClCCl

Specifications

Decomposition Information >120°C
Melting Point -97.0°C
Color Colorless
Density 1.3250g/mL
Boiling Point 39.0°C to 40.0°C
Infrared Spectrum Authentic
Assay Percent Range 99.8% min. (GC)
Packaging AcroSeal™ Glass Bottle
Linear Formula CH2Cl2
Quantity 500 mL
Beilstein 01, 60
Fieser 01,676; 02,273; 04,337; 07,239; 13,106
Specific Gravity 1.325
Merck Index 15, 6135
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: soluble in alcohol, ether and dmf
Stabilizer 50ppm (approx.) amylene
Viscosity 0.43 mPa.s (20°C)
Vapor Pressure 475hPa at 20°C
Formula Weight 84.93
Percent Purity 99.8%
Grade Extra Dry over Molecular Sieve
Physical Form Liquid
Chemical Name or Material Dichloromethane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Suspected of causing cancer.
May cause drowsiness or dizziness.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective cloth
EINECSNumber 200-838-9

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.