Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methoxy-2-tetralone 95.0+%, TCI America™
SDP

Catalog No. M15431G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M15431G 1 g
M15435G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M15431G Supplier TCI America Supplier No. M15431G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 32940-15-1
Molecular Formula C11H12O2
Molecular Weight (g/mol) 176.22
MDL Number MFCD00064958
InChI Key MDAIAXRTLTVEOU-UHFFFAOYSA-N
Synonym 3,4-Dihydro-5-methoxy-2(1H)-naphthalenone
PubChem CID 259194
IUPAC Name 5-methoxy-1,2,3,4-tetrahydronaphthalen-2-one
SMILES COC1=CC=CC2=C1CCC(=O)C2

Specifications

Melting Point 37°C
Color White-Yellow
Boiling Point 165°C
Quantity 1 g
Formula Weight 176.22
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Methoxy-2-tetralone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.