Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-furanmethanol, 97%

Catalog No. ac358510050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

CAS: 3857-25-8 | C6H8O2 | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3857-25-8
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
InChI Key VOZFDEJGHQWZHU-UHFFFAOYSA-N
Synonym 5-methylfuran-2-yl methanol, 5-methyl-2-furanmethanol, 5-methylfurfuryl alcohol, 5-methyl-2-furyl methanol, 2-furanmethanol, 5-methyl, unii-0jvo5efa0a, 0jvo5efa0a, pubchem9804, 5-methyl furfuryl alcohol, 5-methylfuran-2-methanol
PubChem CID 520911
IUPAC Name (5-methylfuran-2-yl)methanol
SMILES CC1=CC=C(O1)CO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Yellow
Density 1.0769g/mL
Boiling Point 80°C to 80.5°C (123.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4835 to 1.4855
Quantity 5 g
Beilstein 17, V,3, 387
Specific Gravity 1.0769
Solubility Information Solubility in water: insoluble
Formula Weight 112.13
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-furanmethanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.