Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-Methyl-2-thiopheneboronic Acid (contains varying amounts of Anhydride), TCI America™

Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Chemical Name or Material | 5-Methyl-2-thiopheneboronic Acid (contains varying amounts of Anhydride) |
CAS | 162607-20-7 |
Melting Point | 150°C |
Color | Yellow |
Molecular Formula | C5H7BO2S |
MDL Number | MFCD01318166 |
Quantity | 1 g |
Synonym | 5-methylthiophene-2-boronic acid, 5-methylthiophen-2-yl boronic acid, 5-methyl-2-thiopheneboronic acid, 5-methyl-2-thienyl boronic acid, 5-methyl-2-thienylboric acid, 5-methylthiophen-2-yl boranediol, 5-methyl-2-thienylboronic acid, 5-methylthiophene-2-boronicacid, 2-methylthiophene-5-boronic acid, 2-methylthienyl-5-boronic acid |
InChI Key | NRIYPIBRPGAWDD-UHFFFAOYSA-N |
SMILES | CC1=CC=C(S1)B(O)O |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction