Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-2-thiophenecarboxylic acid, 99%, Thermo Scientific™

Catalog No. AC127880500
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1918-79-2
MDL Number MFCD00005439
InChI Key VCNGNQLPFHVODE-UHFFFAOYSA-N
Synonym 5-methyl-2-thiophenecarboxylic acid, 5-methyl-2-thenoic acid, 2-thiophenecarboxylic acid, 5-methyl, 2-carboxy-5-methylthiophene, 5-methly-2-thiophenecarboxylic acid, 5-methyl-thiophene-2-carboxylic acid, 5-methyl-2-thiophene carboxylic acid, 5-methylthiophen-2-carboxylic acid, 2-methyl-5-thiophenecarboxylic acid, pubchem5206
PubChem CID 74713
IUPAC Name 5-methylthiophene-2-carboxylic acid
SMILES CC1=CC=C(S1)C(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 136°C to 139°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C6H6O2S
Quantity 50 g
Beilstein 18, 295
Molecular Weight (g/mol) 142.172
Formula Weight 142.18
Percent Purity 98.5 to 101.5% (Acid)
Physical Form Liquid
Chemical Name or Material 5-Methyl-2-thiophenecarboxylic acid
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.