Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic Acid Hydrochloride 97.0+%, TCI America™
SDP

Catalog No. M28191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M28191G 1 g
M2819200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M28191G Supplier TCI America Supplier No. M28191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 720720-96-7
Molecular Formula C8H11ClN2O2S
Molecular Weight (g/mol) 234.698
MDL Number MFCD25542379
InChI Key ZWIYEBIMFPQYDI-UHFFFAOYSA-N
PubChem CID 11149031
IUPAC Name 5-methyl-6,7-dihydro-4H-[1,3]thiazolo[5,4-c]pyridine-2-carboxylic acid;hydrochloride
SMILES CN1CCC2=C(C1)SC(=N2)C(=O)O.Cl

Specifications

Melting Point 201°C
Color Yellow
Quantity 1 g
Formula Weight 234.70
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Methyl-4,5,6,7-tetrahydrothiazolo[5,4-c]pyridine-2-carboxylic Acid Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.