Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylcytosine (5-mC) Monoclonal Antibody (A1), Active Motif™
SDP

Catalog No. 501993433
Encompass
Change view
Click to view available options
Quantity:
10 μg
100 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-3433 10 μg
50-199-3432 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-3433 Supplier Active Motif Supplier No. 61480
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Methylation of DNA cytosine bases plays important roles in the regulation of gene transcription. It has been estimated that 60 to 90% of the cytosines in CpG dinucleotides are methylated. An increase in methyl-CpG correlates with transcriptional silencing for the whole chromosome especially in developmentally regulated genes. Methylation is believed to lead to transcriptional silencing by multiple mechanisms including alteration of transcription factor binding to promoters and alteration in chromatin structure.

Specifications

Antigen 5-Methylcytosine (5-mC)
Applications ChIP sequencing (ChIP-seq), Dot Blot, Immunoprecipitation
Classification Monoclonal
Clone A1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Gene Alias 5mc; 5-mC; 5-meC; 5-Methyl-cytosine; C5H7NO3
Host Species Mouse
Immunogen This 5-Methylcytosine (5-mC) antibody was raised against 5-Methyl-cytidine conjugated to kLH and recognizes 5-Methylcytosine.
Purification Method Protein A
Quantity 10 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.