Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Methylcytosine (5-mC) Mouse anti-Human, Clone: 33D3, Active Motif™
SDP

Catalog No. 501992759
Encompass
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-2759 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-199-2759 Supplier Active Motif Supplier No. 39649
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Methylation of DNA cytosine bases plays important roles in the regulation of gene transcription. It has been estimated that 60 to 90% of the cytosines in CpG dinucleotides are methylated. An increase in methyl-CpG correlates with transcriptional silencing for the whole chromosome especially in developmentally regulated genes. Methylation is believed to lead to transcriptional silencing by multiple mechanisms including alteration of transcription factor binding to promoters and alteration in chromatin structure.

Specifications

Antigen 5-Methylcytosine (5-mC)
Applications Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 33D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 10mM phosphate with 0.15M NaCl and 0.01% thimerosal; pH 7.5
Gene Alias 5mc; 5-mC; 5-meC; 5-Methyl-cytosine; C5H7NO3
Host Species Mouse
Immunogen Clone 33D3 recognizes the modified base 5-methylcytidine found in plant and vertebrate DNA.
Purification Method Protein A
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.