Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5-methylcytosine Recombinant Mouse Monoclonal Antibody, AbFlex™, Active Motif™

Mouse Recombinant Monoclonal Antibody
Supplier: Active Motif 91187

Description
Methylation of DNA cytosine bases plays important roles in the regulation of gene transcription. It has been estimated that 60 to 90% of the cytosines in CpG dinucleotides are methylated. An increase in methyl-CpG correlates with transcriptional silencing for the whole chromosome especially in developmentally regulated genes. Methylation is believed to lead to transcriptional silencing by multiple mechanisms including alteration of transcription factor binding to promoters and alteration in chromatin structure.Specifications
5-methylcytosine | |
Monoclonal | |
Unconjugated | |
5mc; 5-mC; 5-meC; 5-Methyl-cytosine; C5H7NO3 | |
This antibody was raised against 5-methylcytosine conjugated to kLH. | |
100 μg | |
Primary | |
Antibody | |
IgG2a |
Dot Blot | |
1 mg/mL | |
140mM HEPES, 35mM sodium acetate with 30% glycerol, 70mM NaCl and 0.035% sodium azide; pH 7.5 | |
Mouse | |
Protein A | |
RUO | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid | |
Chemical |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction