Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(5-Methylisoxazol-3-yl)methanol, 97%, Thermo Scientific™

Catalog No. cc10409da
Click to view available options
Quantity:
1 g
5 g
10 g

Chemical Identifiers

CAS 35166-33-7
Molecular Formula C5H7NO2
Molecular Weight (g/mol) 113.12
MDL Number MFCD00085129
InChI Key MDYHWQQHEWDJKR-UHFFFAOYSA-N
Synonym 5-methylisoxazol-3-yl methanol, 3-hydroxymethyl-5-methylisoxazole, 5-methyl-1,2-oxazol-3-yl methanol, 5-methyl-3-isoxazolemethanol, 5-methyl-isoxazol-3-yl-methanol, 5-methylisoxazole-3-methanol, 5-methyl-3-isoxazolyl methanol, 5-methyl-isoxazol-3-yl methanol, 3-isoxazolemethanol, 5-methyl
PubChem CID 2736567
IUPAC Name (5-methyl-1,2-oxazol-3-yl)methanol
SMILES CC1=CC(CO)=NO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 113.12
Percent Purity 97%
Chemical Name or Material (5-Methylisoxazol-3-yl)methanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.