Learn More
Greiner Bio-One 5 mL Sterile Serological Pipets
Click to view available options
Packaging Type:
Bulk
Individual Paper/Plastic Wrap
Individual Plastic Wrap
Description
- High-grade polystyrene
- Clear, easily legible graduation
- Colored vertical stripe for ease in reading the volume
- With synthetic fibrous filter material for protection against
Specifications
Specifications
Material | Polystyrene |
Sterility | Sterile |
Graduated | Yes |
Graduations | 1/10 |
Packaging Type | Individual Paper/Plastic Wrap |
Volume (Metric) | 5 mL |
Type | Serological Pipet |