Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitro-2-(3-phenylpropylamino)benzoic acid, 98%

Catalog No. p-7057684
Click to view available options
Quantity:
10 mg
50 mg

Catalog No. AAJ67152MA

Supplier: Thermo Scientific Chemicals J67152MA

Only null left
Add to Cart

CAS: 107254-86-4 | C16H16N2O4 | 300.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107254-86-4
Molecular Formula C16H16N2O4
Molecular Weight (g/mol) 300.31
MDL Number MFCD00153851
InChI Key WBSMIPAMAXNXFS-UHFFFAOYSA-N
Synonym nppb, 5-nitro-2-3-phenylpropylamino benzoic acid, unii-3a35o9g3yz, 5-nitro-2-3-phenylpropyl amino benzoic acid, benzoic acid,5-nitro-2-3-phenylpropyl amino, benzoic acid, 5-nitro-2-3-phenylpropyl amino, 5-nitro-2-3-phenylpropylamino-benzoic acid, 5-nitro-2-phenylpropylaminobenzoic acid nppb, 5-nitro-2-3-phenylpropylamino benzoic acid nppb, spectrum_001814
PubChem CID 4549
ChEBI CHEBI:34457
IUPAC Name 5-nitro-2-[(3-phenylpropyl)amino]benzoic acid
SMILES OC(=O)C1=CC(=CC=C1NCCCC1=CC=CC=C1)[N+]([O-])=O

Specifications

Melting Point 178°C to 180°C
Color Yellow
Quantity 10 mg
Merck Index 14,6725
Formula Weight 300.31
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 5-Nitro-2-(3-phenylpropylamino)benzoic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.