Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitro-2-furaldehyde, 99%

Catalog No. p-7080882
Click to view available options
Quantity:
25 g
50 g

CAS: 698-63-5 | C5H3NO4 | 141.082 g/mol

5-Nitro-2-furaldehyde was used in the synthesis of a series of 4-(5-nitrofuran-2-yl)prop-2-en-1-one derivatives. . It was also used in the synthesis of modified mesoporous silica (MCM-41). 5-Nitrofurfural can be commonly used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-2-furaldehyde was used in the synthesis of a series of 4-(5-nitrofuran-2-yl)prop-2-en-1-one derivatives. . It was also used in the synthesis of modified mesoporous silica (MCM-41). 5-Nitrofurfural can be commonly used as pharmaceutical intermediates.

Solubility
Sparingly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 698-63-5
Molecular Formula C5H3NO4
MDL Number MFCD00003230
InChI Key SXINBFXPADXIEY-UHFFFAOYSA-N
Synonym 5-nitro-2-furaldehyde, 5-nitrofurfural, nitrofurfural, 2-furancarboxaldehyde, 5-nitro, 5-nitrofurfuraldehyde, 5-nitro-2-furfural, 2-furaldehyde, 5-nitro, 5-nitrofuraldehyde, 5-nitro-2-furancarboxaldehyde, furfural, 5-nitro
PubChem CID 12774
ChEBI CHEBI:34459
IUPAC Name 5-nitrofuran-2-carbaldehyde

Specifications

Melting Point 34°C to 38°C
Density 1.349
Boiling Point 228°C
Flash Point 33°C (91°F)
Refractive Index 1.59
Quantity 25 g
Beilstein 120539
Sensitivity Air and light sensitive
Solubility Information Sparingly soluble in water.
SMILES C1=C(OC(=C1)[N+](=O)[O-])C=O
Molecular Weight (g/mol) 141.082
Formula Weight 141.09
Percent Purity 99%
Chemical Name or Material 5-Nitro-2-furaldehyde
Show More Show Less
Hazard Category H228
Hazard Statement GHS H Statement
H341
Suspected of causing genetic defects.
Precautionary Statement P210-P240-P241-P280-P370+P378q
DOTInformation Hazard Class: 4.1; Packaging Group: III
EINECSNumber 211-816-3
RTECSNumber LT7200000
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.