Learn More
5-Nitro-2-furaldehyde semicarbazone, 98+%
Description
Bactericidal compound used as an antibiotic
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∼240°C (decomposition) |
| Quantity | 100 g |
| Beilstein | 86403 |
| Sensitivity | Light sensitive |
| Merck Index | 14,6600 |
| Formula Weight | 198.14 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 5-Nitro-2-furaldehyde semicarbazone |
Safety and Handling
| Hazard Category | H302-H317 |
| Hazard Statement | GHS H Statement H301-H341-H351-H361 Toxic if swallowed. Suspected of causing genetic defects. Suspected of causing cancer. Suspected of damaging fertility or the unborn child. |
| Precautionary Statement | P261-P264b-P270-P272-P280g-P301+P312-P302+P352-P330-P333+P313-P363-P501c |
| EINECSNumber | 200-443-1 |
| RTECSNumber | LT7700000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.