Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitroacenaphthene (contains 3-Nitroacenaphthene) 80.0+%, TCI America™
SDP

Catalog No. N010525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N010525G 25 g
N01055G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N010525G Supplier TCI America Supplier No. N010525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 602-87-9
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.21
MDL Number MFCD00014277
InChI Key CUARLQDWYSRQDF-UHFFFAOYSA-N
Synonym 5-nitroacenaphthene, 5-nitroacenapthene, acenaphthene, 5-nitro, acenaphthylene, 1,2-dihydro-5-nitro, 5-nan, 1,2-dihydro-5-nitro-acenaphthylene, 5-nitronaphthalene ethylene, 5-nitronaphthalene, 1,2-dihydro-5-nitroacenaphthylene, ccris 438
PubChem CID 11769
ChEBI CHEBI:82350
IUPAC Name 5-nitro-1,2-dihydroacenaphthylene
SMILES [O-][N+](=O)C1=C2C=CC=C3CCC(C=C1)=C23

Specifications

Melting Point 102°C
Color Yellow
Quantity 25 g
UN Number 2811
Formula Weight 199.21
Percent Purity ≥80.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Nitroacenaphthene (contains 3-Nitroacenaphthene)
EINECSNumber (4)-0352
RTECSNumber AB1060000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.