Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Nitrobenzimidazole, 98+%

Catalog No. p-7080815
Click to view available options
Quantity:
50 g
250 g

CAS: 94-52-0 | C7H5N3O2 | 163.14 g/mol

5-Nitrobenzimidazole is used as sensitive reagents determinating of bismuth, cadmium, copper,and rubber antioxidant. It also used as photographic materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitrobenzimidazole is used as sensitive reagents determinating of bismuth, cadmium, copper,and rubber antioxidant. It also used as photographic materials.

Solubility
Sparingly soluble in water. (0.5g/L at 25°C). Soluble in methanol.

Notes
Incompatible with strong oxidizing agents, strong bases. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-52-0
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.14
MDL Number MFCD00005604
InChI Key XPAZGLFMMUODDK-UHFFFAOYSA-N
Synonym 5-nitrobenzimidazole, 6-nitrobenzimidazole, 5-nitro-1h-benzo d imidazole, 5-nitro-1h-benzimidazole, 1h-benzimidazole, 5-nitro, 1h-benzimidazole, 6-nitro, 6-nitro-benzimidazole, 5 6-nitrobenzimidazole, benzimidazole, 5-nitro, benzimidazole, 6-nitro
PubChem CID 7195
IUPAC Name 6-nitro-1H-benzimidazole
SMILES [O-][N+](=O)C1=CC=C2N=CNC2=C1

Specifications

Melting Point 207°C to 211°C
Quantity 50 g
Beilstein 7926
Solubility Information Sparingly soluble in water. (0.5g/L at 25°C). Soluble in methanol.
Formula Weight 163.14
Percent Purity ≥98%
Chemical Name or Material 5-Nitrobenzimidazole
Hazard Category H302-H351
Hazard Statement GHS H Statement
H351
Suspected of causing cancer.
Precautionary Statement P201-P202-P264b-P270-P281-P301+P312-P308+P313-P330-P501c
EINECSNumber 202-341-2
RTECSNumber DD9800000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.