Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 5-Nitroindole-2-carboxylic acid, 96%

Click to view available options
Quantity:
1g
5g
16730-20-4
C9H6N2O4
206.157
MFCD00236372
LHFOJSCXLFKDIR-UHFFFAOYSA-N
5-nitroindole-2-carboxylic acid,5-nitro-1h-indole-2-carboxylic acid,1h-indole-2-carboxylic acid, 5-nitro,5-nitro-2-indolecarboxylic acid,1h-indole-2-carboxylicacid, 5-nitro,5-nitroindole-2-carboxylicacid,pubchem1710,zlchem 1340,5-nitroindole-2-carboxylic acid,5-nitro indole-2-carboxylic acid
351291
C1=CC2=C(C=C1[N+](=O)[O-])C=C(N2)C(=O)O
16730-20-4
C9H6N2O4
206.157
MFCD00236372
LHFOJSCXLFKDIR-UHFFFAOYSA-N
5-nitroindole-2-carboxylic acid,5-nitro-1h-indole-2-carboxylic acid,1h-indole-2-carboxylic acid, 5-nitro,5-nitro-2-indolecarboxylic acid,1h-indole-2-carboxylicacid, 5-nitro,5-nitroindole-2-carboxylicacid,pubchem1710,zlchem 1340,5-nitroindole-2-carboxylic acid,5-nitro indole-2-carboxylic acid
351291
C1=CC2=C(C=C1[N+](=O)[O-])C=C(N2)C(=O)O
Quantity 5g
Beilstein 203744
Formula Weight 206.16
Physical Form Solid
Percent Purity 96%
Chemical Name or Material 5-Nitroindole-2-carboxylic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning