Alfa Aesar™ 5-Nitroindole, 99%

Manufacturer: Alfa Aesar™ A1060803

Quantity 1g

 View more versions of this product

Catalog No. AAA1060803

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 6146-52-7
    Molecular Formula C8H6N2O2
    Formula Weight 162.15
    MDL Number MFCD00005673
    Beilstein 383777
    Chemical Name or Material 5-Nitroindole

    Specifications

    Formula Weight 162.15
    Health Hazard 3 GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H341
    Suspected of causing genetic defects.
    Molecular Formula C8H6N2O2
    MDL Number MFCD00005673
    CAS 6146-52-7
    RTECS Number NM1168200
    Melting Point 139° to 143°C
    Quantity 1g
    Beilstein 383777
    Health Hazard 1 Warning
    Chemical Name or Material 5-Nitroindole
    Assay 99%

    GHS H Statement
    H341
    Suspected of causing genetic defects.

    GHS P Statement
    P281-P201-P202-P308+P313-P405-P501a
    Use personal protective equipment as required.
    Obtain special instructions before use.
    Do not handle until all safety precautions have been read and understood.
    IF exposed or concerned: Get medical advice/attention.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit