Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Norbornene-2-carboxamide (mixture of isomers) 98.0+%, TCI America™
SDP

Catalog No. N091125G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 95-17-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00187896
InChI Key ZTUUVDYQBLRAAC-UHFFFAOYSA-N
Synonym Bicyclo[2.2.1]hept-5-ene-2-carboxamide
PubChem CID 96112
IUPAC Name bicyclo[2.2.1]hept-2-ene-5-carboxamide
SMILES C1C2CC(C1C=C2)C(=O)N

Specifications

Melting Point 188°C
Color White-Yellow
Quantity 25 g
Formula Weight 137.18
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Norbornene-2-carboxamide (mixture of isomers)
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.