Learn More
5'-Nucleotidase/CD73 Antibody (45M4F9), Alexa Fluor™ 350, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP225243AF350
Catalog No. NP22243A30
Only null left
Add to Cart
Description
Description
5'-Nucleotidase/CD73 Monoclonal specifically detects 5'-Nucleotidase/CD73 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
5′-Nucleotidase/CD73
Monoclonal
Alexa Fluor 350
50 mM sodium borate with 0.05% sodium azide
NT5E
Within the range of amino acids 169-324 of human CD73 were used as the immunogen for the antibody.
Primary
This product can detect Isoform 1 and Isoform of Human CD73.
Store at 4°C in the dark.
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
45M4F9
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
5′ nucleotidase (CD73), 5′-NT, 5′-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5′-nucleotidase, eN, eNT, NT, NT55′-nucleotidase, NTE, Purine 5-Prime-Nucleotidase
Mouse
0.1 mL
4907
Human
IgG2b κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.