Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5'-Nucleotidase/CD73 Antibody (496406), DyLight 680, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB4488Y
Description
5'-Nucleotidase/CD73 Monoclonal antibody specifically detects 5'-Nucleotidase/CD73 in Mouse samples. It is validated for Flow Cytometry, CyTOFSpecifications
5'-Nucleotidase/CD73 | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, CyTOF | |
496406 | |
Flow Cytometry, CyTOF-ready | |
5' nucleotidase (CD73), 5'-NT, 5'-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5'-nucleotidase, eN, eNT, NT, NT55'-nucleotidase, NTE, Purine 5-Prime-Nucleotidase | |
Chinese hamster ovary cell line CHO-derived recombinant mouse 5'-Nucleotidase/CD73, Trp29-Lys549, Accession # Q61503 | |
0.1 mL | |
Lipid and Metabolism, Myeloid derived Suppressor Cell | |
4907 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction