Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
5'-Nucleotidase/CD73 Antibody (OTI1G2), FITC, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271214F
Description
5'-Nucleotidase/CD73 Monoclonal specifically detects 5'-Nucleotidase/CD73 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.Specifications
5′-Nucleotidase/CD73 | |
Monoclonal | |
FITC | |
PBS with 0.05% Sodium Azide | |
NT5E | |
Recombinant protein fragment corresponding to amino acids 119-232 of human 5′-Nucleotidase/CD73 (NP_002517) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Immunohistochemistry | |
OTI1G2 | |
Immunohistochemistry | |
5′ nucleotidase (CD73), 5′-NT, 5′-nucleotidase, ecto (CD73), CD73, CD73 antigen, E5NT, EC 3.1.3.5, ecto-5′-nucleotidase, eN, eNT, NT, NT55′-nucleotidase, NTE, Purine 5-Prime-Nucleotidase | |
Mouse | |
Affinity Purified | |
Lipid and Metabolism, Myeloid derived Suppressor Cell | |
4907.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction