Learn More
Description
To enquire about custom knock-in cell lines for the AID2 platform, please email TPD@bio-techne.com
Plasmid vectors for the AAV transfection and CRISPR-mediated knock-in of OsTIR1F74G and mAID tag are available from Addgene .
Specifications
Specifications
| Chemical Name or Material | 5-Phenyl-1 H-indole-3-acetic acid |
| CAS | 168649-23-8 |
| Quantity | 10 mg |
| Target | Active Degraders |
| Molecular Formula | C16H13NO2 |
| Purity | 0.98 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

